Learn More
MSC Honeywell First Responder Products Rig Dog™ Xtreme Mechanics Gloves, Impact Resistant, Cut-Resistant, 10/XL, 6/Pk.
Supplier: MSC 15003817
These RigDog Series mechanics gloves are engineered to provide impact protection and enhanced grip in demanding work environments. Designed for versatility, they feature specialized construction for cold resistance and are ideal for handling oily surfaces while ensuring operator comfort.
- Reinforced with thermoplastic rubber pads on the fingers and back of the hand for impact protection
- Constructed with microfiber for superior abrasion resistance and oil grip
- Lined with a winter insert for use in cold environments
- Features a knitted inner layer for improved comfort and sweat absorption
- Includes a hook-and-loop closure for a secure fit
- Highly breathable design for extended wear