Learn More
Medchemexpress LLC Poly(styrene)-block-poly(acrylic acid) (PS:PAA 3,000:5,000,DDMAT terminated) | 10 MG
SDP
Supplier:  Medchemexpress LLC 10MGHY172378
Encompass_Preferred
Poly(styrene)-block-poly(acrylic acid) (PS:PAA 3,000:5,000,DDMAT terminated) is a diblock copolymer used to prepare polymer micelles or vesicles and other encapsulation applications.
  • Diblock copolymer
  • Used to prepare polymer micelles or vesicles
  • Suitable for encapsulation applications
Catalog No. 50-003-65307