Learn More
Medchemexpress LLC Poly(styrene)-block-poly(acrylic acid) (PS:PAA 30000:2000) | 90.0% | 32000 | 5 MG
Supplier: Medchemexpress LLC 5MGHY172378
Poly(styrene)-block-poly(acrylic acid) (PS:PAA 30000:2000) is a diblock copolymer primarily used for preparing polymer micelles, vesicles, and other encapsulation applications. It is intended for research use only and not for human or animal therapeutic use. It appears as a solid with a color ranging from white to off-white, and can be stored at room temperature in powder form. It is soluble in DMF and THF, requiring ultrasonic assistance for dissolution.
- Used to prepare polymer micelles or vesicles
- Suitable for encapsulation applications
- Appears as a solid, white to off-white
- Soluble in DMF (25 mg/mL) and THF (20 mg/mL) with ultrasonic assistance
- Stable as powder at -20°C for 3 years