Learn More
Medchemexpress LLC Poly(styrene)-block-poly(acrylic acid) (PS:PAA 30000:2000) | 90.0% | 50 MG

Supplier: Medchemexpress LLC 50MGHY172378
Poly(styrene)-block-poly(acrylic acid) (PS:PAA 30000:2000) is a diblock copolymer utilized in the preparation of polymer micelles or vesicles and other encapsulation applications. This product is intended for research use only.
- Solid appearance
- White to off-white color
- Soluble in DMF (25 mg/mL with ultrasonic) and THF (20 mg/mL with ultrasonic)
- Ships at room temperature in continental US
- Storage as powder at -20°C for 3 years
- Storage in solvent at -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.