Learn More
Sigma Aldrich Fine Chemicals Biosciences Poly(vinyl alcohol), 9002-89-5, MFCD00081922, 250 G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences P8136250G
Encompass
Linear Formula: [-CH2CHOH-]n, 87-90% hydrolyzed, average mol wt 30,000-70,000. Poly(vinyl alcohol) has been used: in in vitro production (IVP) of bovine embryos, oocyte recovery and in vitro maturation, and as a macromolecular component in basal cultured medium.
Catalog No. NC1216859