Learn More
Medchemexpress LLC Poly(vinyl alcohol) | 9002-89-5 | MFCD00081922 | 92.0-94.0% | 146,000-186,000 | (C2H4O)n | 25g
SDP
Supplier:  Medchemexpress LLC HYY0850I25G
Encompass_Preferred
Polyvinyl alcohol (Mw 9000-10000 80% hydrolyzed) also known as PVA is a vinyl water-soluble polymer that can be used as a non-ionic surfactant Polyvinyl alcohol (Mw 9000-10000 80% hydrolyzed) can be used as a biodegradable polymer and can be used in adhesives coatings textiles ceramics and cosmetics Polyvinyl alcohol can be used in tissue engineering by electrospinning Polyvinyl alcohol can achieve high cellular density infiltration and uniform distribution facilitating functional connections between cells Polyvinyl alcohol can improve cell vitality through in vitro cultivation Polyvinyl alcohol demonstrates promising inhibition of ostersarcoma cancer cells with Doxorubicin (HY-15142A)[1][2]
Catalog No. 50-002-69753