Learn More
MSC Miller Titan II Full Body Harness, Polyester, S/M, 1/ea.
Supplier: MSC 65899544
This high-visibility full body harness is designed for general construction and industrial applications. It features a lightweight, non-stretch polyester construction and a sliding back D-ring for proper dorsal alignment and improved user comfort. The harness provides five points of adjustment and is rated for a working load capacity of 400 lbs.
- Lightweight, non-stretch polyester webbing
- High-visibility orange color for increased safety
- Sliding back D-ring for optimal alignment
- Five points of adjustment for a secure fit
- Mating buckle chest straps and tongue-buckle leg straps
- Meets OSHA, ANSI, and CSA standards