Learn More
Medchemexpress LLC Polyacrylamide Anion 25g
SDP
Supplier:  Medchemexpress LLC 25GHYW115727A
Encompass_Preferred
Polyacrylamide Anion Mw 14-16 million is a multifunctional high molecular weight anionic polyacrylamide copolymer The anionic properties of Polyacrylamide Anion Mw 14-16 million enable it to be used as a flocculant to achieve charge neutralization and aggregation while its high molecular weight properties provide viscoelastic properties for fluid applications Polyacrylamide series materials can maintain enzyme activity in enzyme immobilization achieve controlled release as a drug carrier respond to temperature/pH stimulation as a smart material and can also be used for in vitro toxin adsorption and soft tissue filling through mechanisms such as physical embedding covalent bonding or chemical cross-linking Polyacrylamide can be used in biomedical engineering environmental management and industrial applications[1][2]
Catalog No. 50-002-04641