Learn More
Medchemexpress LLC Polyaniline | 25233-30-1 | MFCD00241446 | 98.6% | Polymeric (variable); repeat unit molar mass 93.13 g/mol | (C6H7N)x | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW017767
Encompass_Preferred
Polyaniline is a conductive polymer used in research and materials science, including development of antibacterial agents and electronic materials. Supplied as a powder, the material is polymeric with variable molecular weight and is reported at 98.6% purity, making it suitable for synthesis, coatings, and analytical studies.
  • Conductive polymer for antibacterial agent development and materials research.
  • Supplied as a powder suitable for synthesis, coating, and characterization.
  • Reported purity 98.6% supports reproducible experimental results.
  • Polymeric nature with variable molecular weight; determine grade-specific Mw for applications.
  • Dispersible in common laboratory solvents for processing and testing.
Catalog No. 50-003-04212