Learn More
Medchemexpress LLC Polycarbophil | 9003-97-8 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1546591G
Encompass_Preferred
Polycarbophil is an orally active hydrophilic polymer. It achieves bioadhesion through hydrogen bonding between carboxyl groups and mucosal surfaces. It prolongs compound retention time and regulates compound release through its swelling properties. It is used to relieve constipation and diarrhea.
  • Orally active hydrophilic polymer
  • Achieves bioadhesion through hydrogen bonding with mucosal surfaces
  • Prolongs compound retention time
  • Regulates compound release through swelling properties
  • Relieves constipation and diarrhea
  • Increases apparent permeability of compounds (in vitro)
  • Increases fecal weight and water content (in vivo)
  • Restores fecal weight and relieves constipation (in vivo)
  • Suppresses diarrhea (in vivo)
Catalog No. 50-003-76892