Learn More
Medchemexpress LLC Polycarbophil | 9003-97-8 | [Polycarbophil] | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY154659
Encompass_Preferred
Polycarbophil is an orally active hydrophilic polymer. It achieves bioadhesion through hydrogen bonding between carboxyl groups and mucosal surfaces, prolonging compound retention time and regulating compound release through swelling properties.
  • Used to relieve both constipation and diarrhea.
  • Increases apparent permeability of Pipemidic acid in isolated porcine urinary bladder wall.
  • Increases fecal weight and water content in naive rats.
  • Restores fecal weight and relieves constipation in constipated rats.
  • Reduces stress-induced fecal output.
  • Suppresses Castor oil-induced diarrhea in rats.
  • For research use only.
Catalog No. 50-003-75944