Learn More
Vector Laboratories Polyclonal Rabbit anti‑Human Complement C1s Antibody (WB) LS‑C748341, 100ul
SDP
Supplier:  Vector Laboratories LSC748341100
Encompass
Complement C1s antibody LS-C748341 is an unconjugated rabbit polyclonal antibody to Complement C1s from human. It is reactive with human, mouse and rat. Validated for WB.
Catalog No. NC3804766