Learn More
Medchemexpress LLC Polycytidylic acid potassium | 26936-40-3 | MFCD00131956 | 99.2% | (polymeric; molecular weight not specified) | (C9H14N3O8P)x.xK | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW25015250MG
Encompass_Preferred
Polycytidylic acid potassium is a synthetic double-stranded RNA used experimentally to model viral dsRNA and activate innate immune responses via TLR3. It is supplied as a solid material intended for research applications in immunology and vaccine adjuvant studies.
  • High purity suitable for research use (99.18%).
  • Synthetic double-stranded RNA that acts as a TLR3 agonist.
  • Useful for immune activation, antiviral response modeling, and adjuvant studies.
  • White to off-white solid form, easy to weigh and handle.
  • Storage guidance: store sealed, away from moisture; in solvent: -80°C for long-term, -20°C short-term.
Catalog No. 50-002-81301