Learn More
Advanced Biomatrix Poly-D-Lysine, Solution, 0.1 mg/mL
Catalog No. NC0876312
Encompass_Preferred
Poly-D-Lysine comes with 50 mL of a 0.1 mg/ml sterile-filtered solution. Poly-D-Lysine is a synthetic amino acid chain that is positively charged and widely used as a coating to enhance cell attachment and adhesion to both plasticware and glass surfaces. This molecule is resistant to enzymatic degradation and has been used to culture a wide variety of cell types. The molecular weight of Poly-D-Lysine can vary significantly with lower molecular weight (30,000 Da) being less viscous and higher molecular weight (>300,000 Da) having more binding sites per molecule. This product’s molecular weight ranges from 70,000 to 150,000 Da yielding a solution viscosity for easy handling while providing sufficient binding sites for cell attachment. Poly-D-Lysine surface coatings are designed to improve cell attachment, growth and differentiation of many cell types. Coated surfaces will often improve cell attachment in reduced or serum-free conditions.
Catalog No. NC0876312 Supplier Advanced Biomatrix Supplier No. 504950ML
Poly-D-Lysine comes with 50 mL of a 0.1 mg/ml sterile-filtered solution. Poly-D-Lysine is a synthetic amino acid chain that is positively charged and widely used as a coating to enhance cell attachment and adhesion to both plasticware and glass surfaces. This molecule is resistant to enzymatic degradation and has been used to culture a wide variety of cell types. The molecular weight of Poly-D-Lysine can vary significantly with lower molecular weight (30,000 Da) being less viscous and higher molecular weight (>300,000 Da) having more binding sites per molecule. This product’s molecular weight ranges from 70,000 to 150,000 Da yielding a solution viscosity for easy handling while providing sufficient binding sites for cell attachment. Poly-D-Lysine surface coatings are designed to improve cell attachment, growth and differentiation of many cell types. Coated surfaces will often improve cell attachment in reduced or serum-free conditions.