Learn More
U.S. Pharmacopeia Polydimethylsiloxane, 9016-00-6, MFCD00148360, 500mg
Supplier:  U.S. Pharmacopeia 1546300
Encompass
Molecular formula (C2H6OSi)n, Molecular weight 71.8 - 547.1 kg/mol, Melting Point -58 °F (-50 °C), Boiling Point > 600.8 °F (> 316 °C), Flash point 384.8 °F (196.0 °C) (closed cup), Synonyms: Silicone oil, Dimethylsiloxane, Dimethicone 350
Catalog No. NC9617656