Learn More
Cole-Parmer Polyethylene Tape, 1" W; 36 yd/RL, 12 RL/Cs
Supplier:  Cole-Parmer 0827910
Encompass_Preferred
Excellent for sealing or resealing jar lids. White tape stretches and conforms to wrap flasks, vacuum bottles, and other glass vessels. Use to reduce injury risk from implosion.
Catalog No. NC1778402