Learn More
Braintree Scientific Polyethylene .034"""" x .060"""" Per Ft.,100 ft. Continuous
SDP
Supplier:  Braintree Scientific PE100100FT
Encompass
Polyethylene Tubing Polyethylene is a relatively rigid polymer suitable for short-term catheterizations and for ex vivo applications. Polyethylene sleeves can be slipped over less rigid catheter materials (MicroRenathane, RenaSil™) at suture points to protect the latter from inadvertent constriction **Sold for research purposes only and not for human use**
Catalog No. 50-195-5488
 Restricted Item