Learn More
Medchemexpress LLC Polyethylene glycol 1000 | 25322-68-3 | MFCD01779605 | 1000.00 | H(OCH2CH2)nOH | 100g
SDP
Supplier:  Medchemexpress LLC HYY0873G100G
Encompass_Preferred
PEG1000 can be used as an excipient such as Ointment base lubricant etc Pharmaceutical excipients or pharmaceutical auxiliaries refer to other chemical substances used in the pharmaceutical process other than pharmaceutical ingredients Pharmaceutical excipients generally refer to inactive ingredients in pharmaceutical preparations which can improve the stability solubility and processability of pharmaceutical preparations Pharmaceutical excipients also affect the absorption distribution metabolism and elimination (ADME) processes of co-administered drugs[1]
Catalog No. 50-002-68538