Learn More
Kyfora Bio Polyethylene glycol 1450
Supplier:  Kyfora Bio 00679250
Encompass_Preferred
Polyethylene glycol (PEG) is a water-soluble biologically inert polymer that has a number of biological and pharmaceutical applications Specifically PEG1450 can be used as an excipient in pharmaceutical products or as a coating on nanoparticles to prevent opsonization in vivo It can also be used for purification of plasmid DNA The polymer chains are hydroxyl terminated at both ends
Catalog No. 50-002-85306