Learn More
Medchemexpress LLC Polyethylene glycol 3000 | 25322-68-3 | 98.0% | 3000.00 g/mol | H(OCH2CH2)nOH | 100g
SDP
Supplier:  Medchemexpress LLC HYY0873N100G
Encompass_Preferred
PEG3000 (Polyethylene glycol 3000) is a solvent for a large number of substances PEG3000 can be used as a carrier material and modifying agent PEG3000 is widely used in a variety of pharmaceutical formulations[1]
Catalog No. 50-002-68564