Learn More
Crescent Chemical Co Inc Polyethylene Glycol 40 000, 25322-68-3, MFCD00081839, 500g
SDP
Supplier:  Crescent Chemical Co Inc SE33139.01
Encompass_Preferred
Polyethylene Glycol 40 000, 25322-68-3, MFCD00081839, 500g, Research Grade, Ph. Eur., USP, Degree of polymerization ca. 800 - 900.Polyethylene glycol for chromatography, histology, microscopy and for special biochemical purposes.
Catalog No. 50-248-035