Learn More
Crescent Chemical Co Inc POLYETHYLENE GLYCOL 40 000
SDP
Supplier:  Crescent Chemical Co Inc SE33139.02
Encompass_Preferred
Polyethylene glycol 40 000 research grade, Ph. Eur., USP Degree of polymerization ca. 800 - 900.Polyethylene glycol for chromatography, histology, microscopy and for special biochemical purposes. Size - 5KG Storage Conditions - +15 °C TO +30 °C Catalog Number
Catalog No. 50-248-036