Learn More
Medchemexpress LLC Polyethylene glycol 8000 | 25322-68-3 | MFCD00081839 | 99.9% | 8000.00 | (C2H4O)n H2O | 25g
SDP
Supplier:  Medchemexpress LLC HYY0873J25G
Encompass_Preferred
PEG8000 can be used for phage precipitation isolation of plasmid DNA and promotion of flat end-junction reactions[1]
Catalog No. 50-002-68779