Learn More
Sigma Aldrich Fine Chemicals Biosciences Polyethylene glycol diamin5G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 7530841G
Encompass
Poly(ethylene glycol) diamine (PEGD) is a non-toxic biopolymer which has properties such as biocompatibility and biodegradability. It can be reacted with crosslinking agents to form a three dimensional mesh that can be used in drug delivery systems. It can also be used as a non-ionic surfactant for a variety of biological applications.
Catalog No. 50-290-6746