Learn More
Kyfora Bio PEI MAX - Transfection Grade (Linear Polyethylenimine Hydrochloride MW 40,000) 1GR
Supplier:  Kyfora Bio 247651
Encompass_Preferred
Kyfora Bio’s Polyethyleneimine MAX (PEI MAX) is a powerful, reliable, and cost-effective reagent, widely regarded as the gold standard for both in vitro and in vivo applications. PEI MAX can be used to produce high viral, recombinant protein, and monoclonal antibody titers across a diverse array of cell lines, including HEK293 and CHO cells. It offers exceptional transfection efficiency for both stable and transient transfections in adherent and suspension cultures, all while preserving cell viability, making it an ideal transfection reagent compared to other products on the market.
Catalog No. 50-255-9822