Learn More
Kyfora Bio Polyethylenimine, Linear, MW 250,000 (LPEI 250K) 2GR
Supplier:  Kyfora Bio 243142
Encompass
Linear polyethylenimine (L-PEI 250,000) is a high-charge cationic polymer that readily binds highly anionic substrates. In research, PEI readily binds to DNA and other negatively charged biological molecules. It can be used for electrostatic layer-by-layer deposition of plasmid DNA for gene delivery.Key application(s): Gene delivery
Catalog No. 50-255-9823