Learn More
Medchemexpress LLC Polygalacic acid | 22338-71-2 | 504.70 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN08015MG
Encompass_Preferred
Polygalacic acid is a triterpene isolated from the root of *Polygala tenuifolia Willd*. It inhibits matrix metalloproteinase (MMP) expression and may offer a therapeutic effect in Osteoarthritis (OA) treatment. It also demonstrates a significant neuroprotective effect on cognitive impairment by improving cholinergic system reactivity, achieved through inhibiting acetylcholinesterase (AChE) activity, increasing choline acetyltransferase (ChAT) activity, and elevating acetylcholine (Ach) levels in the hippocampus and frontal cortex.
  • Inhibits matrix metalloproteinase (MMP) expression.
  • May have therapeutic effect in Osteoarthritis (OA) treatment.
  • Exerts neuroprotective effect on cognitive impairment.
  • Improves cholinergic system reactivity.
  • Inhibits acetylcholinesterase (AChE) activity.
  • Increases choline acetyltransferase (ChAT) activity.
  • Elevates acetylcholine (Ach) levels.
Catalog No. 50-004-15304