Learn More
Medchemexpress LLC Polygalaxanthone III | 162857-78-5 | MFCD23704367 | 568.48 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN14071MG
Encompass_Preferred
Polygalaxanthone III is extracted from polygala tenuifolia wild and has an inhibitory effect towards the CYP450 enzyme. It inhibits chlorzoxazone 6-hydroxylation catalyzed by CYP2E1 with an IC50 of 50.56 μM.
  • Extracted from polygala tenuifolia wild
  • Inhibits cyp450 enzyme
  • Inhibits chlorzoxazone 6-hydroxylation catalyzed by cyp2e1
  • For research use only
Catalog No. 50-004-15762