Learn More
Cayman Chemical Polygalaxnthon III 5mg
SDP
Supplier:  Cayman Chemical 306215 MG
Encompass_Preferred
A xanthone glycoside with antioxidant activity; scavenges hydroxyl and DPPH radicals in cell-free assays (IC50s = 83.5 and 76.1 µM, respectively)
Catalog No. 50-299-0772