Learn More
Enzo Life Sciences Polygodial (25mg). CAS: 6754-20-7
SDP
Supplier:  Enzo Life Sciences BMLCA2420025
Encompass_Preferred
Produced from the bark of Drymis winteri. Naturally occurring drimane sesquiterpene found in water pepper (Polygonum hydropiper) which has been used in folk medicine for a variety of indications. Potent and selective activator of TRPA1 (EC50= 400nM). Alternative name: (1R,4aS,8aS)-1,4,4a,5,6,7,8,8a-Octahydro-5,5,8a-trimethyl-1,2-naphthalenedicarboxaldehyde. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (25 mg/ml) or ethanol (25 mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0381