Learn More
Nanocs Polylysine, Rhodamine labeled, 2mg
Supplier:  Nanocs PL1RB1
Encompass
Molecular weight: ~25 kD, ~100 lysine units. Other MW available upon request.2~7 functional groups on each poly-L-lysine molecule.Polylysine conjugates provided either in water solution or dried powder;
Catalog No. NC2681823