Learn More
United States Biological Corporation Polymannuronic Acid, Sodium Salt CAS: 29894-36-8
Supplier:  United States Biological Corporation 2976451G
Encompass_Preferred
Sodium polymannuronate is produced from alginates by partial hydrolysis and chromatography of brown algae such as Laminaria digitata, Ascophyllum nodosum and Fucus spp. Food additive; anti-tumoral.
Catalog No. NC3836228