Learn More
Fritsch Milling And Sizing Inc Complete Polymer Coating Of The Inside Of The Grinding Chamber, P-13 Pl
SDP
Supplier:  Fritsch Milling And Sizing Inc 13.3090.16
Encompass
Ideal for medical and dental engineering: On request, we can equip your PULVERISETTE 13 premium line with a complete polymer interior coating together with grinding disks made of zirconium oxide – the perfect prerequisite for grinding without any contact of the sample with metal.
Catalog No. 50-153-3940
Only null left
Add to Cart
Request a Quote