Learn More
Medchemexpress LLC Polymethyl methacrylate | 9011-14-7 | MFCD00134349 | (C5H8O2)n | 100 G
SDP
Supplier:  Medchemexpress LLC HYW391506100G
Encompass_Preferred
Poly(methyl methacrylate) (PMMA) beads are a transparent, rigid acrylic polymer used for fabrication of microfluidic chips and other laboratory applications that require an optically clear, stable thermoplastic. Supplied as beads for easy handling and processing, this material is suitable for molding, machining, and prototyping in research settings. For research use only; not for human or clinical applications.
  • Transparent, rigid acrylic polymer suitable for optical applications.
  • Supplied as beads for easy handling and accurate dosing.
  • Suitable for microfluidic chip fabrication and prototyping.
  • Compatible with common machining, molding, and casting processes.
  • Insoluble in water and stable under standard laboratory conditions.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-85288