Learn More
Crescent Chemical Co Inc POLYMYXIN B-SULFATE
SDP
Supplier:  Crescent Chemical Co Inc SE47976.03
Encompass_Preferred
Polymyxin-B-sulfate research grade, Ph. Eur. Mixture of the sulfates of polypeptides produced by the growth of certain strains of Bacillus polymyxa, the main component being Polymyxin B1.Assay (HPLC): sum of Polymyxins B1, B2, B3 and B1-I min. 80 %; Polymyxin B3 max.
Catalog No. 50-249-142