Learn More
Medchemexpress LLC Polyoxyethylene stearate | 9004-99-3 | MFCD00148007 | 328.53 g/mol | C20H40O3 | 10 G
SDP
Supplier:  Medchemexpress LLC HY101530C10G
Encompass_Preferred
Polyoxyethylene stearate is a non-ionic emulsifying agent (PEG-100 stearate) commonly used in formulations and laboratory research to stabilize oil-in-water systems, act as a solubilizer, and improve texture and spreadability. It is supplied as a white to off-white solid (CAS 9004-99-3) with a monomer molecular weight around 328.5 g/mol.
  • White to off-white solid suitable for formulation use.
  • Non-ionic emulsifier for oil-in-water systems.
  • Acts as a solubilizer and surfactant to improve dispersion.
  • Helps stabilize emulsions and improve texture.
  • Compatible with a wide pH range and common excipients.
  • Available in small research pack sizes for laboratory use.
Catalog No. 50-002-48362