Learn More
U.S. Pharmacopeia Polyoxyl 35 Castor Oil, 61791-12-6, MFCD00130879, 1g
Supplier:  U.S. Pharmacopeia 1547200
Encompass
Dynamic viscosity 1318.51 mPa.s (73.4 °F (23 °C)), Flash point > 212.0 °F (> 100.0 °C), Chemical name: Polyoxyethylenglycerol triricinoleate 35
Catalog No. NC1655756