Learn More
Medchemexpress LLC Polyoxypropylene stearyl ether | 25231-21-4 | (C3H6O)nC18H38O | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY154630
Encompass_Preferred
Polyoxypropylene stearyl ether can be used as an excipient, functioning as a surfactant, softener, lubricating, wetting, plasticizing, solubilizing, and dispersing agent. Pharmaceutical excipients are inactive ingredients in pharmaceutical preparations that improve stability, solubility, and processability, and can also affect the absorption, distribution, metabolism, and elimination (ADME) processes of co-administered drugs.
  • Functions as a surfactant
  • Acts as a softener and lubricating agent
  • Used for wetting and plasticizing
  • Aids in solubilizing and dispersing
  • Improves stability and solubility in pharmaceutical preparations
  • Enhances processability of formulations
  • Influences drug absorption, distribution, metabolism, and elimination
Catalog No. 50-003-75943