Learn More
Sigma Aldrich Fine Chemicals Biosciences PolypepR Low Viscosity his100G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences P5115100G
Encompass
A proprietary protein digest that provides increased stablization of tissue sections in histochemical studies.
Catalog No. 50-290-4953