Learn More
Apexbio Technology LLC Polyphyllin A 50773-41-6 20mg
SDP
Supplier:  Apexbio Technology LLC N192420
Encompass_Preferred
Polyphyllin A (CAS 50773-41-6) is a steroidal saponin isolated from the rhizomes of Paris yunnanensis Franch It is known to modulate multiple cellular signaling pathways including the induction of apoptosis and inhibition of proliferation in various cancer cell lines Polyphyllin A exerts its biological activity by affecting mitochondrial membrane potential promoting cytochrome c release and activating caspase cascades Due to these properties it serves as a valuable tool for investigating cell death mechanisms and evaluating potential anticancer strategies in preclinical biomedical research The compound is soluble in methanol and ethanol and should be stored sealed in a cool dry environment
Catalog No. 50-000-94177