Learn More
Medchemexpress LLC Polyporusterone B | 141360-89-6 | 5 MG

Supplier: Medchemexpress LLC HYN76935MG
Polyporusterone B is a triterpene carboxylic acid isolated from Polyporus umbellatus Fries. It demonstrates an inhibitory effect on the free radical-induced lysis of red blood cells, also known as hemolysis. This compound is intended for research use only.
- Triterpene carboxylic acid.
- Isolated from Polyporus umbellatus Fries.
- Inhibits free radical-induced lysis of red blood cells.
- Research use only.
- Purity: 99.24%.
- Appearance: Solid.
- Color: White to off-white.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.