Learn More
Elemental Scientific Inc POLYPRO-ST NEBULIZER
SDP
Supplier:  Elemental Scientific Inc ES4040
Encompass_Preferred
PolyPro ST nebulizer. Peristaltic pumped or self-aspirated high performance nebulizer for ICP and ICPMS. Includes two capillaries (100ul/min. & 400ul/min.)
Catalog No. NC0692145