Learn More
Andwin Scientific POLYPROPYLENE DEEP WELL MICROP
SDP
Supplier:  Andwin Scientific 40002012
Encompass_Preferred
The 96-well deep well microplates can work with both automated and robotic systems. This product is also compatible to use cell harvesters and many microplate centrifuge rotors. The plates are resistant against chemicals and are ideal for high-throughput screening applications, including drug discovery and diagnostic research. This product is also alphanumerically labeled that allows convenient and easy sample identification. Each square well has a volume capacity of 2.0 mL and a conical bottom.
Catalog No. NC0063080