Learn More
AirSea USA 50ml Biotube with 19mm bung – suitable for the shipment of Class 6.2 infectious and biological substances, Category A UN 2814 and UN 2900 and Category B UN 3373, when used within an approved outer packaging.
Supplier:  AirSea USA 334
Encompass_Preferred
50ml Biotube with 19mm bung – suitable for the shipment of Class 6.2 infectious and biological substances when used within an approved outer packaging. Approved with Code 333 (Temperature Control Packaging, Class 6.2 Infectious and Biological Substances) and Code 474
Catalog No. NC2101116