Learn More
Alkali Scientific Reusable Glass Volumetric Pipets, Class A, Color Coded, Colored Markings, 10/Pk, 3mL, Black, ±0.015mL Tolerance, 360mm
SDP
Supplier:  Alkali Scientific GVF003
Encompass
Reusable Glass Volumetric Pipets are precision manufactured Class A transfer pipettes designed for accurate and repeatable liquid measurement in analytical, research, and quality control laboratories. Each pipet features a single calibration mark with permanent color coded markings that allow quick volume identification while reducing reading errors. Manufactured from high quality laboratory glass, these pipets meet strict Class A accuracy tolerances and provide controlled outflow times for consistent dispensing. Their reusable design supports long term laboratory use while maintaining the precision required for critical procedures such as solution preparation, titrations, and reference measurements. Available in volumes ranging from 1 mL to 100 mL, each is color coded and supplied 10 pipets per pack for convenient laboratory use.
Catalog No. 50-006-61917