Learn More
TWD Scientific LLC Extra Small Polypropylene Weighing Funnels. Pack of 96
Supplier: TWD Scientific LLC DPWFPPXS
Disposable polypropylene weighing funnel, size extra small. Fits 96-well microtubes.Funnels have a maximum capacity of 1mL Solid/0.5mL Liquid. 96/Pack