Learn More
Cayman Chemical PolysarcosIn50 10mg
SDP
Supplier:  Cayman Chemical 3904910 MG
Encompass_Preferred
An amine-functionalized hydrophilic polymer; has been used in the synthesis of catechol-functionalized copolypeptoids that can be used to solubilize manganese oxide nanoparticles in aqueous solvents; surface modification of liposomes with polysarcosine polymers between 10 and 400 units long have been used to increase circulation half-life
Catalog No. 50-291-8497