Learn More
U.S. Pharmacopeia Polysorbate 80, 9005-65-6, MFCD00082107, 4 x 0.5 g
Supplier:  U.S. Pharmacopeia 1547969
Encompass
Boiling Point 212 °F (100 °C), Flash point 298.4 °F (148.0 °C) Closed Cup, Synonyms: Polyoxyethylene 20 sorbitan monooleate * Glycol
Catalog No. NC9304824