Learn More
Polysciences, Inc. Polybead Polystyrene Microsphere, Monodisperse Latex Beads, 1um, 1.00um Particles
SDP
Supplier:  Polysciences, Inc. 0731015
Encompass
Polybead Polystyrene Microsphere, Monodisperse Latex Beads, 1um, 1.00um Particles Packaged as 2.5% Aqueous Suspension, BP: 100 deg.C, MP: 0 deg.C, Opaque Liquid, Store at 4 deg.C
Catalog No. NC9862439