Learn More
Medchemexpress LLC Polyvinyl alcohol | 9002-89-5 | 13,000-23,000 g/mol | (C2H4O)x | 5g
SDP
Supplier:  Medchemexpress LLC HYY0850J5G
Encompass_Preferred
Polyvinyl alcohol (Mw 13000-23000 87-89% hydrolyzed) is a polyvinyl alcohol with a molecular weight of 130000-23000 and hydrolytic properties Polyvinyl alcohol (Mw 13000-23000 87-89% hydrolyzed) is the hydrolysis and removal of acetate groups after the polymerization of ethylene acetate and polyvinyl alcohol is obtained A degree of hydrolysis of 87-89% indicates that a large part of the acetate groups have been removed resulting in a large number of hydroxyl groups in the PVA structure Polyvinyl alcohol with different degrees of hydrolysis can be used to self-crosslink to form cryogel which can be used as biological excipient Polyvinyl alcohol can be used in tissue engineering by electrospinning Polyvinyl alcohol can achieve high cellular density infiltration and uniform distribution facilitating functional connections between cells Polyvinyl alcohol can improve cell vitality through in vitro cultivation Polyvinyl alcohol demonstrates promising inhibition of ost
Catalog No. 50-002-68504